Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG F(ab')2 fragment,Polyclonal Secondary Antibody, Abnova™

Catalog No. 89139021 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-021 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-021 Supplier Abnova Corporation Supplier No. PAB9355
Add to Cart
Add to Cart

Specifications

Antigen Goat anti-Mouse IgG F(ab')2 fragment
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat anti-mouse IgG recognizes mouse IgG F(ab')2 fragment. This secondary antibody was purified using delipidation and defibrination.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM KH2PO4, 150mM NaCl, pH 7.2
Host Species Goat
Immunogen Mouse IgG F(ab')2 fragment
Purification Method This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against mouse IgG F(ab')2 and mouse serum.
Quantity 2 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.