Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Mouse, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ303FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ303FMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ303FMI Supplier MilliporeSigma Supplier No. AQ303F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

This Goat anti-Mouse IgG Antibody, F(ab′)2, FITC conjugate is validated for use in IF for the detection of Mouse IgG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgG
Applications Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate FITC
Formulation Liquid in 0.02M Phosphate Buffer. 0.25M NaCl, pH 7.6 with 15mg/mL BSA and 0.01% Sodium azide.
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for upto 12 months.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.