Learn More
Goat anti-Mouse IgG F(c) fragment, Rhodamine, TRITC, Polyclonal, Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Goat anti-Mouse IgG F(c) fragment |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Rhodamine |
| Description | Goat anti-mouse IgG recognizes mouse IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Rhodamine. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) sodium azide, 10mg/mL BSA (ImmunoglobulLyophilized from and Protease free)) |
| Host Species | Goat |
| Immunogen | Mouse IgG F(c) fragment |
| Purification Method | This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.