Learn More
Arbor Assays Goat Anti-Mouse IgG Fc, 10MG
SDP
Supplier:  Arbor Assays A00810MG
Encompass_Preferred
An affinity purified Goat Antibody to Mouse IgG, Fc region. Supplied in PBS with azide as preservative. Affinity purified on a column with covalently coupled mouse IgG, Fc attached.
Catalog No. 50-227-8555