Learn More
Invitrogen™ Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody, Biotin
Description
Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody for ELISA, Flow, ICC/IF, Western Blot
Specifications
Specifications
| Antigen | Mouse IgG Fc Cross-Adsorbed |
| Applications | ELISA, Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 1% BSA and 0.05% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Gamma Immunoglobin Fc region. |
| Purification Method | Affinity chromatography |
| Quantity | 2 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.