Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP127PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP127PMI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP127PMI Supplier MilliporeSigma Supplier No. AP127P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Detect Mouse IgG using this Goat anti-Mouse IgG Antibody, flow cytometry, HRP conjugate validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8mg/mL
Conjugate HRP
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA. Contains no preservative. Reconstitute with sterile distilled water to match the volume indicated on the vial label.
Host Species Goat
Quantity 2 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for 6 months
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.