Learn More
Invitrogen™ Goat anti-Mouse IgG Fc Secondary Antibody, FITC
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Specifications
Specifications
| Antigen | Mouse IgG Fc |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 1% BSA and 0.05% sodium azide; pH 7.8 |
| Host Species | Goat |
| Immunogen | Gamma Immunoglobin Fc region. |
| Purification Method | Affinity chromatography |
| Quantity | 2 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.