Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP181MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP181MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP181MI Supplier MilliporeSigma Supplier No. AP181
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Goat anti-Mouse IgG Antibody, Species Adsorbed detects level of Mouse IgG and has been published and validated for use in ELISA, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgG Fc Species adsorbed
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Goat
Quantity 1 mg
Primary or Secondary Secondary
Target Species Mouse
Form Liquid

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.