Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Mouse IgG Fc Superclonal™ Secondary Antibody, Alexa Fluor™ Plus 647

Catalog No. 5000031030
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-000-31030 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-000-31030 Supplier Invitrogen™ Supplier No. SA510374AFP647
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Recombinant Superclonal Secondary Antibody

Goat anti-Mouse IgG Fc, Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ Plus 647 specifically detects the Fc portion of mouse IgG1, mouse IgG2a, and mouse IgG2b with no reactivity to Mouse IgG F(ab)2 fragment and with minimum cross reactivity to Human IgG, Rat IgG and Hamster IgG. The sensitivity and specificity of each lot of Goat anti-Mouse IgG Fc, Superclonal™ Recombinant Secondary Antibody is confirmed using ELISA and then taken up for conjugation. Superclonal™ recombinant secondary antibodies are produced using a multifaceted clonal selection process that involves several phenotypic screens to ensure enhanced specificity, animal origin free formulation and lot-to-lot consistency. Alexa Fluor™ Plus conjugated recombinant antibodies provide bright signals and high signal-to-noise for your critical experiments. It is recommended that the antibody is carefully titrated for optimal performance in the assay of interest.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG Fc
Applications Peptide Array, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Superclonal
Concentration 1.0 mg/mL
Conjugate Alexa Fluor Plus 647
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.3-7.3
Host Species Goat
Immunogen Purified Mouse IgG.
Purification Method Affinity chromatography
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.