Learn More
Jackson Immuno Research Labs Fluorescein (FITC) AffiniPure Goat Anti-Mouse IgG, Fcγ fragment specific, 1.5mg
SDP
Supplier:  Jackson Immuno Research Labs 115095071
Encompass_Preferred
Fluorescein (FITC)-AffiniPure Goat Anti-Mouse IgG, Fc Fragment Specific (min X Hu,Bov,Hrs Sr Prot)
Catalog No. NC9484818