Learn More
Jackson Immuno Research Labs Biotin-SP (long spacer) AffiniPure Goat Anti-Mouse IgG (H+L), .5ml
SDP
Catalog No. NC9372582
Encompass_Preferred
Biotin-SP-AffiniPure Goat Anti-Mouse IgG (H+L) (min X Rat,Hu,Bov,Hrs,Rb Sr Prot)
Catalog No. NC9372582 Supplier Jackson Immuno Research Labs Supplier No. 115065166
Biotin-SP-AffiniPure Goat Anti-Mouse IgG (H+L) (min X Rat,Hu,Bov,Hrs,Rb Sr Prot)