Learn More
MilliporeSigma™ Goat anti- Mouse IgG, H + L Chain Specific, Peroxidase, Secondary Antibody,
Shop All MilliporeSigma ProductsDescription
Specifically detects IgG, H&L Chain Specific Clone: in Mouse samples, and it is validated for Enzyme Immunoassay, Immunoelectrophoresis
Specifications
Specifications
| Antigen | IgG, H&L Chain Specific |
| Applications | ELISA, Immunoelectrophoresis |
| Classification | Polyclonal |
| Conjugate | Peroxidase |
| Formulation | In 10mM PBS, 0.5mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6. |
| Host Species | Goat |
| Quantity | 2 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Test Specificity | Adsorbed against bovine, horse, and human serum proteins. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.