Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Mouse IgG, H + L Chain Specific, Secondary Antibody,

Catalog No. 4012111ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
40-121-11ML 1 mL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-121-11ML Supplier MilliporeSigma™ Supplier No. 4012111ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Specifically detects IgG, H & L Chain Specific Clone: in Mouse samples, and it is validated for Immunoelectrophoresis, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG, H & L Chain Specific
Applications Immunoelectrophoresis, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In 10mM PBS, pH 8.0.
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage −20°C
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.