Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (H+L) Goat anti-Mouse, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 12506MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-506-MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-506-MI Supplier MilliporeSigma Supplier No. 12506
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Detect Mouse IgG (H+L) using this Goat anti-Mouse IgG (H+L) Antibody, FITC conjugate validated for use in flow cytometry, immunofluoresence, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgG (HL)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate FITC
Formulation Lyophilized powder; lyophilized from 250μl of 20mM potassium phosphatase, pH 7.2, 150mM NaCl, 0.01% sodium azide and 10mg/mL BSA.
Host Species Goat
Immunogen Mouse IgG whole molecule.
Purification Method Affinity Purified
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.