Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP308PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP308PMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP308PMI Supplier MilliporeSigma Supplier No. AP308P
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Goat anti-Mouse IgG Antibody, (H+L) HRP conjugate is validated for use in ELISA, immunocytochemistry, immunohistochemistry, western blotting for the detection of Mouse IgG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgG
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Purified Goat IgG in buffer containing liquid in PBS, pH 7.1 with 15mg/mL BSA and 0.01% thimerosal.
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C in undiluted format for 12 months. Do not Freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.