Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, bGal
Description
Goat anti-Mouse IgG (H+L) Secondary Antibody for ELISA
Specifications
Specifications
| Antigen | Mouse IgG (H+L) |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | b-GAL (beta-Galactosidase) |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide |
| Host Species | Goat |
| Immunogen | Proprietary Immunogen. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.