Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG heavy and light chains, FITC, Polyclonal, Secondary Antibody, Abnova™

Catalog No. 89136878 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-878 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-878 Supplier Abnova Corporation Supplier No. PAB0103
Add to Cart
Add to Cart

Specifications

Antigen Goat anti-Mouse IgG heavy and light chains
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate FITC
Description Goat anti-mouse IgG recognizes mouse IgG heavy and light chains. This secondary antibody was purified using antigen affinity chromatography and adsorbed against human proteins to minimize species cross-reactivity. The purified antibodies are then conjugated to Fluorescein (FITC), a small fluorescent organic molecule with a molecular weight of 473 daltons with an absorbance maximum of ∼492 nm and an emission maximum of ∼518 nm.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide, 1.0% BSA)
Host Species Goat
Immunogen Purified Mouse IgG whole molecule (>98 %)
Purification Method This product was purified using antigen affinity chromatography and adsorbed against serum proteins and immunoglobulins to minimize species cross-reactivity.
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C. Do not Freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.