Learn More
Cell Signaling Technology Goat Anti-Mouse IgG (H+L) Antibody, Alexa Fluor™ 750 Conjugate, 250 μL
SDP
Catalog No. 503133325
Encompass
This goat anti-mouse IgG (H+L) antibody is conjugated to Alexa Fluor™ 750 fluorescent dye and is designed for use in immunofluorescent applications. This secondary antibody conjugate detects the heavy and light chains of mouse IgG and has been cross-adsorbed against human IgG and human serum for increased specificity.
  • Optimized for flow cytometry and immunofluorescence applications
  • Supplied at a concentration of 2 mg/mL
  • Excitation maximum at 749 nm and emission maximum at 775 nm
  • Validated for use in immunohistochemistry and flow cytometry assays
  • Formulated in 0.1 M sodium phosphate, 0.1 M sodium chloride, pH 7.5, and 5 mM sodium azide
  • Recommended dilution of 1:500 to 1:2,000 for flow cytometry and 1:125 for paraffin-embedded tissues
Catalog No. 50-313-3325 Supplier Cell Signaling Technology Supplier No. 67939S
This goat anti-mouse IgG (H+L) antibody is conjugated to Alexa Fluor™ 750 fluorescent dye and is designed for use in immunofluorescent applications. This secondary antibody conjugate detects the heavy and light chains of mouse IgG and has been cross-adsorbed against human IgG and human serum for increased specificity.
  • Optimized for flow cytometry and immunofluorescence applications
  • Supplied at a concentration of 2 mg/mL
  • Excitation maximum at 749 nm and emission maximum at 775 nm
  • Validated for use in immunohistochemistry and flow cytometry assays
  • Formulated in 0.1 M sodium phosphate, 0.1 M sodium chloride, pH 7.5, and 5 mM sodium azide
  • Recommended dilution of 1:500 to 1:2,000 for flow cytometry and 1:125 for paraffin-embedded tissues