Learn More
Enzo Life Sciences Goat anti-mouse IgG, (HRP conjugate) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA2040100
Encompass_Preferred
Immunogen: Mouse IgG. Species reactivity: Mouse . Specificity: Recognizes mouse IgG heavy and light chains (whole molecule). Applications: ELISA, WB . Purity: Affinity purified. Formulation: Liquid. In 20 mM potassium phosphate, pH 7.2, containing 150mM sodium chloride, 0.01% gentamicin, and 1% BSA as stabilizer.
Catalog No. 50-201-1768