Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG & IgM Goat anti-Mouse, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP130FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP130FMI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP130FMI Supplier MilliporeSigma Supplier No. AP130F
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Detect Mouse IgG and IgM using this Goat anti-Mouse IgG and IgM Antibody, FITC conjugate validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgG IgM
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Host Species Goat
Quantity 2 mg
Primary or Secondary Secondary
Target Species Mouse

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.