Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Mouse IgG, IgM (H+L) Secondary Antibody, AP

Catalog No. T2192
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2192 Supplier Invitrogen™ Supplier No. T2192
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Secondary Antibody

This Alkaline Phosphatase Conjugated secondary antibody can be used with the ELISA-Light Immunoassay, Western-Light, and Western-Star Western Blotting Detection Systems. This labeled secondary antibody ensures minimal nonspecific cross-reactivity. Thermo Scientific has optimized its alkaline phosphatase secondary antibody conjugates for Western blotting or immunoassay procedures incorporating chemiluminescent 1,2-dioxetane enzyme substrates. Optimized for use in blotting applications with dioxetane-based substrate solutions (CSPD or CDP-Star substrates), providing consistent experiment-to-experiment results. High specific activity of conjugates provides high sensitivity of detection with dioxetane-based substrate solutions (CSPD or CDP-Star substrates). Low non-specific binding provides highest possible signal-to-noise in western blotting and immunoassay applications. Easy to Use The recommended initial working dilution for the labeled antibodies is 1:5,000 for AP conjugates. The conjugate is tested and optimized for low background with chemiluminescent detection procedures.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG, IgM (H+L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation TBS with 4-5% BSA, 40% glycerol and 0.05% sodium azide
Host Species Goat
Immunogen Mouse IgG/IgM.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.