Learn More
Invitrogen™ Goat anti-Mouse IgG, IgM, IgA (H+L) Secondary Antibody, Alexa Fluor™ 488
Description
Product will be shipped at Room Temperature.
Specifications
Specifications
| Antigen | Mouse IgG, IgM, IgA (H+L) |
| Applications | Flow Cytometry, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Formulation | PBS with 5mM sodium azide; pH 7.5 |
| Host Species | Goat |
| Immunogen | Mouse IgG/IgA/IgM (H+L). |
| Purification Method | Affinity chromatography |
| Quantity | 500 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.