Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioLegend™ Goat anti-mouse IgG , Secondary

Catalog No. NC9049740
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC9049740 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC9049740 Supplier BioLegend™ Supplier No. 405307
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Host Species: Mouse
Clone: Poly4053
Isotype: IgG
Cross Reactivity: Goat

Flow Cytometry

Specifications

Antigen Goat anti-mouse IgG
Classification Monoclonal
Clone Poly4053
Concentration 0.2mg/mL
Conjugate PE
Host Species Mouse
Purification Method Unpurified
Quantity 200 μg
Primary or Secondary Secondary
Target Species Goat
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.