Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioLegend™ Goat anti-mouse IgG , Secondary

Catalog No. NC9664552
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC9664552 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC9664552 Supplier BioLegend™ Supplier No. 405306
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Host Species: Goat
Clone: Poly4053
Isotype: IgG
Cross Reactivity: Mouse

ELISA, Immunohistochemistry, Western Blotting

Specifications

Antigen Goat anti-mouse IgG
Classification Monoclonal
Clone Poly4053
Conjugate HRP
Host Species Goat
Purification Method Unpurified
Quantity 500 μL
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.