Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ127MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ127MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ127MI Supplier MilliporeSigma Supplier No. AQ127
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Goat anti-Mouse IgG Antibody, Fc detects level of Mouse IgG and has been published and validated for use in ELISA, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgG Species adsorbed
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity Purified F(ab')2 Antibody in liquid buffer of 0.01% Sodium Phosphate, 0.25M NaCl, pH 7.6 containing no preservatives.
Host Species Goat
Immunogen purified mouse IgG Fc fragment
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.