Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Mouse IgG1/2a/2b Fc, Clone: RMG06, Secondary Antibody,

Catalog No. 041662MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
04-166-2MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-166-2MI Supplier MilliporeSigma™ Supplier No. 041662
Add to Cart
Add to Cart

Goat Monoclonal Antibody

Specifically detects IgG1/2a/2b Fc Clone: RMG06 in Mouse samples, and it is validated for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1/2a/2b Fc
Applications ELISA
Classification Monoclonal
Clone RMG06
Conjugate Unconjugated
Formulation Goat monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Host Species Goat
Immunogen Mouse IgG
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary, Control Antibodies
Primary or Secondary Secondary
Test Specificity This antibody reacts to the Fc region of Mouse IgG, including IgG1, IgG2a, and IgG2b. It shows very slight reaction with mouse IgG3. No cross reactivity with IgM, IgA, IgE, human IgG, rat IgG, or rabbit IgG.
Target Species Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.