Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Mouse IgG1 Fc Cross-Adsorbed Secondary Antibody, APC

Catalog No. PI31982 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
300 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31982 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31982 Supplier Invitrogen™ Supplier No. 31982
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31982 reacts with the heavy chains of mouse IgG1, but not with heavy chains of other mouse IgG subclasses or with light chains of most mouse immunoglobulins. This antibody does not react against mouse IgM, or against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, bovine, and rabbit serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Phycobiliprotein Concentration: 0.5 mg/mL (determined by absorption = 73 at 650 nm for a 1% solution for only those APC molecules to which at least one molecule of active antibody is bound) Phycoerythrin Source: Spirulina platensis Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. After dilution, do not use for more than one day. Do not freeze.Allophycocyanin Amax= 650 nm; Emax= 660 nm. Reconstitute with 0.3 mL of distilled water (0.5 mg/mL after restoration). Country of Origin: USA.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG1 Fc Cross-Adsorbed
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate APC
Formulation PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Host Species Goat
Purification Method Antigen affinity chromatography
Quantity 300 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG Fc
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.