Learn More
Description
This antibody does not react with human. Goat anti Mouse IgG1 antibody recognizes Mouse IgG1.
Specifications
Specifications
| Antigen | Mouse IgG1 |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 50% glycerol and 0.0095% Methylisothiazolone; pH 7.4 |
| Host Species | Goat |
| Immunogen | Mouse IgG1 paraproteins. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.