Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Goat Anti-Mouse IgG2b Antibody, Alexa Fluor™ 488 Conjugate, 250 μl
SDP

Catalog No. 503133340
Encompass

This antibody is conjugated to Alexa Fluor™ 488 fluorescent dye under optimal conditions and is designed for use as a secondary antibody in immunofluorescent applications. This goat-derived antibody specifically detects mouse IgG2b and is suitable for techniques including immunofluorescence (frozen) and immunofluorescence (immunocytochemistry). It is cross-adsorbed to ensure specificity.

  • Excitation: 495 nm; emission: 519 nm.
  • Concentration: 2 mg/mL.
  • Formulated in 0.1 M sodium phosphate, 0.1 M sodium chloride, pH 7.5, and 5 mM sodium azide.
  • Cross-adsorbed against mouse IgM, mouse IgA, pooled human sera, purified human paraproteins, and mouse isotypes IgG1, IgG2a, and IgG3.
  • Recommended final dilution range: 1:500 to 1:2,000.
  • Storage: Store at 4°C. Do not freeze or aliquot. Protect from light.

Catalog No. 50-313-3340 Supplier Cell Signaling Technology Supplier No. 98612S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This antibody is conjugated to Alexa Fluor™ 488 fluorescent dye under optimal conditions and is designed for use as a secondary antibody in immunofluorescent applications. This goat-derived antibody specifically detects mouse IgG2b and is suitable for techniques including immunofluorescence (frozen) and immunofluorescence (immunocytochemistry). It is cross-adsorbed to ensure specificity.

  • Excitation: 495 nm; emission: 519 nm.
  • Concentration: 2 mg/mL.
  • Formulated in 0.1 M sodium phosphate, 0.1 M sodium chloride, pH 7.5, and 5 mM sodium azide.
  • Cross-adsorbed against mouse IgM, mouse IgA, pooled human sera, purified human paraproteins, and mouse isotypes IgG1, IgG2a, and IgG3.
  • Recommended final dilution range: 1:500 to 1:2,000.
  • Storage: Store at 4°C. Do not freeze or aliquot. Protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.