Learn More
Bioworld GOAT ANTI-MOUSE IGGFC)-RE
Supplier:  Bioworld 201810822
Encompass
GOAT ANTI-MOUSE IGG (FC)-RED SEPAROPORE (AGAROSE): Affinity isolated antibody is coupled to cyanogen bromide-activated Separopore4B. The antibody preparation is specific for the Fc fragment of mouse IgG when tested against purified mouse...
Catalog No. 50-197-879