Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Goat anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP128PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP128PMI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP128PMI Supplier MilliporeSigma Supplier No. AP128P
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Goat anti-Mouse IgM Antibody, μ chain, HRP conjugate is validated for use in ELISA, western blotting for the detection of Mouse IgM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgM
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8-1.0mg/mL
Conjugate HRP
Formulation Freeze dried powder, made from liquid containing 0.01M phosphate buffered saline, 0.25M sodium chloride, 15mg/mL bovine serum with no preservatives. Reconstitute in 2mL deionized water, centrifuge if not clear. Prepare a working dilution fresh for each
Host Species Goat
Purification Method Affinity Purified
Quantity 2 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.