Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM μ chain Goat anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP500PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP500PMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP500PMI Supplier MilliporeSigma Supplier No. AP500P
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Detect Mouse IgM μ chain using this Goat anti-Mouse IgM μ chain Antibody, HRP conjugate, Species Adsorbed validated for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgM μ chain Species adsorbed
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation Purified by affinity chromatography on mouse IgM covalently linked to agarose. Liquid in 50% glycerol/50% PBS, pH 7.4.
Host Species Goat
Immunogen purified mouse IgM μ chain. Pooled antisera from goats hyperimmunized with mouse IgM paraproteins.
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C under sterile conditions for up to 12 months −20°C in undiluted aliquots for long term storage. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.