Learn More
Invitrogen™ Goat anti-Mouse IgM Cross-Adsorbed Secondary Antibody, DyLight™ 650
Shop All Thermo Scientific ProductsDescription
This antibody is cross-adsorbed and exhibits minimum reactivity to human and rat.
Specifications
Specifications
| Antigen | Mouse IgM Cross-Adsorbed, Mouse IgM |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | DyLight 650 |
| Formulation | PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 |
| Host Species | Goat |
| Immunogen | Mouse IgM. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 500 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.