Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM+IgG+IgA Goat anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ502PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ502PMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ502PMI Supplier MilliporeSigma Supplier No. AQ502P
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Goat anti-Mouse IgM+IgG+IgA Antibody, F(ab′)2, HRP conjugate, Species Adsorbed is validated for use in ELISA for the detection of Mouse IgM+IgG+IgA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgM IgG IgA F(ab')2 Species adsorbed
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation Purified by gel filtration chromatography of pepsin digested antibody. Liquid in 50% glycerol/50% PBS, pH 7.4.
Host Species Goat
Immunogen pepsin digest of mouse Ig (H+L).
Purification Method Gel filtration chromatography
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C under sterile conditions for up to 12 months −20°C in undiluted aliquots for long term storage. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.