Learn More
MilliporeSigma™ IgM+IgG+IgA (H+L) Goat anti-Mouse, FITC, Polyclonal, Secondary Antibody
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Goat anti-Mouse IgM IgG IgA (HL) Species adsorbed |
| Applications | ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | Purified by affinity chromatography on pooled mouse IgM+IgG+IgA covalently linked to agarose. Liquid in 1.0mLPBS/NaN3. |
| Host Species | Goat |
| Immunogen | pooled antisera from goats hyperimmunized with mouse IgM, IgG and IgA paraproteins. |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Primary or Secondary | Secondary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.