Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM+IgG+IgA (H+L) Goat anti-Mouse, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP501BMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP501BMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP501BMI Supplier MilliporeSigma Supplier No. AP501B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Detect Mouse IgM+IgG+IgA (H+L) using this Goat anti-Mouse IgM+IgG+IgA (H+L) Antibody, biotin conjugate, Species Adsorbed validated for use in ELISA and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse IgM IgG IgA (HL) Species adsorbed
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation Purified by affinity chromatography on pooled mouse IgM+IgG+IgA covalently linked to agarose. Liquid in 2.0mLPBS/NaN3.
Host Species Goat
Immunogen pooled antisera from goats hyperimmunized with mouse IgM, IgG and IgA paraproteins.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C under sterile conditions for up to 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.