Learn More
Description
Product # 31804 reacts with the μ chains of mouse IgM. This antibody cross reacts with Rat IgM (6%). It has minimal cross-reactivity with immunoglobulins from other species.
Specifications
Specifications
| Antigen | Mouse IgM |
| Applications | ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 1mg/mL BSA and 0.08% sodium azide; pH 7.8 |
| Host Species | Goat |
| Purification Method | Affinity chromatography |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.