Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light chain Goat anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP200MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP200MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP200MI Supplier MilliporeSigma Supplier No. AP200
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Detect Mouse light chain using this Goat anti-Mouse light chain Antibody validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse light chain Antibody
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.3mg/mL
Conjugate Unconjugated
Formulation Purified by immunoaffinity chromatography. Liquid in 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6.
Host Species Goat
Immunogen purified mouse IgG light chain.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -20°C under sterile conditions for up to 12 months, Avoid Freeze/Thaw Cycles
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.