Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Pig IgG (H+L) Secondary Antibody, FITC

Catalog No. PIPA184850 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA184850 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA184850 Supplier Invitrogen™ Supplier No. PA184850
Add to Cart
Edge
Add to Cart

Goat Polyclonal Secondary Antibody

Goat anti Pig IgG (H/L) antibody recognizes porcine IgG (heavy and light chains) and has been shown to react specifically with porcine IgG and with light chains common to other pig immunoglobulins, by immunoelectrophoresis and ELISA.

Anti-Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for porcine (swine, pig) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pig IgG (H+L)
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Host Species Goat
Immunogen Proprietary Immunogen.
Purification Method Affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Pig
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.