Learn More
Goat anti-Pig IgG whole molecule, HRP, Polyclonal, Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Goat anti-Pig IgG whole molecule |
| Applications | Dot Blot, ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Description | Goat anti-pig IgG recognizes pig IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) gentamicLyophilized from sulfate (Do NOT add sodium azide!), 10mg/mL BSA (ImmunoglobulLyophilized from and Protease free)) |
| Host Species | Goat |
| Immunogen | Pig IgG whole molecule |
| Purification Method | This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.