Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Rabbit gamma;-Globulin, Secondary Antibody,

Catalog No. 539844125U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
125 U
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
53-984-4125U 125 U Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-984-4125U Supplier MilliporeSigma™ Supplier No. 539844125U
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects gamma;-Globulin Clone: in Rabbit samples, and it is validated for Immunoelectrophoresis, Radioimmunoassay

TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma;-Globulin
Applications Immunoelectrophoresis, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Goat
Quantity 125 U
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity One unit of goat anti-rabbit γ-globulin (GARGG) contains enough antibody to maximally precipitate the γ-globulin in 200μl of 2% normal rabbit serum.
Target Species Rabbit
Content And Storage 2°C to 8°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.