Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat Anti-Rabbit IgG Antibody (H&L) Antibody (HRP), ProSci
SDP

Catalog No. 16838694 Shop All ProSci Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-838-694 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-838-694 Supplier ProSci LLC Supplier No. XG6160HRP0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Secondary Antibody

Goat Anti-Rabbit IgG Antibody specifically detects Goat Anti-Rabbit IgG in Rabbit Samples. It is validated for ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Goat Anti-Rabbit IgG
Applications ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate HRP
Dilution 1:250-1:3,000 for IHC/ICC, , 1:2,000-1:5,000 for ELISA, Western Blot, color substrates., , 1:5,000-1:10,000 for Western Blot, chemiluminescence.
Formulation Buffer containing 0.01M sodium phosphate, 0.25M sodium chloride, pH 7.6 and 25% glycerol.
Host Species Goat
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity This antibody reacts with both heavy and light chains of rabbit IgG, slight cross-reactivity with IgG of other mammalian species.
Target Species Rabbit
Content And Storage Store 2 - 8 °C as an undiluted liquid for several weeks. After dilution, do not use for more than one day. -20 °C for up to one year.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.