Learn More
Enzo Life Sciences Goat anti-rabbit IgG, polyclonal antibody (AP conjugate) (5ml)
SDP
Supplier:  Enzo Life Sciences ENZABS2575000
Encompass_Preferred
This antibody conjugate is used in the AMP'D HSP70 high sensitivity ELISA kit (ENZ-KIT-101) and has been successfully employed for use with the APP ΔC31 ELISA kit (ADI-900-227) as part of our AMP'D detection system to increase the sensitivity of this ELISA ~10-fold. Immunogen: Purified rabbit IgG (whole molecule). Source: Purified from rabbit serum. Species reactivity: Rabbit. Specificity: Recognizes rabbit IgG. Applications: ELISA. Purity: Immunoaffinity purified. Formulation: Liquid (100 ng/ml). In 0.05M TRIS-HCl, pH 8.0, containing 0.075M sodium chloride, 0.05% hydorol M, 0.5% BSA, 0.05% Tween-20, and 50% Stabilzyme-AP.
Catalog No. 50-201-2802