Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Rabbit, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 12448MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-448-MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-448-MI Supplier MilliporeSigma Supplier No. 12448
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

This Goat anti-Rabbit IgG Antibody, Alkaline Phosphatase conjugate is validated for use in ELISA, western blot, immunohistochemistry for the detection of Rabbit IgG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Rabbit IgG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation 50mM Tris chloride, pH 8.0, 150mM NaCl, 1mM MgCl, 0.1mM ZnCl, 0.01% (w/v) sodium azide, 50% (v/v) glycerol and 10mg/mL BSA
Host Species Goat
Quantity 500 μg
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4°C for 12 months. DO NOT FREEZE.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.