Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Rabbit, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP132AMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP132AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP132AMI Supplier MilliporeSigma Supplier No. AP132A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Goat anti-Rabbit IgG Antibody, Alkaline Phosphatase conjugate is an antibody against Rabbit IgG for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Rabbit IgG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6mg/mL
Conjugate Alkaline Phosphatase (AP)
Formulation RECONSTITUTION: Reconstitute with sterile distilled water to match the volume indicated on the vial label. Centrifuge or vortex this product if it is not completely clear after standing for 1-2 hours at room temperature.
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C lyophilized until opened. DO NOT FREEZE.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.