Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Rabbit IgG, Biotin, Secondary Antibody,

Catalog No. OS03B200UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
Conjugate:
Biotin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OS03B200UG 200 μg Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OS03B200UG Supplier MilliporeSigma™ Supplier No. OS03B200UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects IgG Clone: in Rabbit samples, and it is validated for ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation In 250mM NaCl, 10mM sodium phosphate, 15mg/mL BSA, pH 7.6.
Host Species Goat
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage −20°C
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.