Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Rabbit, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP187CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP187CMI 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP187CMI Supplier MilliporeSigma Supplier No. AP187C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

This Goat anti-Rabbit IgG Antibody, Cy3 conjugate, Species Adsorbed is validated for use in IF for the detection of Rabbit IgG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Rabbit IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Concentration 1.5mg/mL
Conjugate Cyanine3
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, 0.01% Thimerosal and 0.05 Sodium Azide. RECONSTITUTION: Reconstitute to 1.5mg/mL with sterile distilled water.
Host Species Goat
Quantity 1.5 mg
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.