Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Rabbit, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ132FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ132FMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ132FMI Supplier MilliporeSigma Supplier No. AQ132F
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Goat anti-Rabbit IgG Antibody, F(ab′)2, FITC conjugate detects level of Rabbit IgG and has been published and validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Rabbit IgG
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Host Species Goat
Quantity 1 mg
Primary or Secondary Secondary
Target Species Rabbit

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.