Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rabbit IgG F(ab')2 fragment, AP, Polyclonal, Secondary Antibody, Abnova™

Catalog No. 89137366 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-366 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-366 Supplier Abnova Corporation Supplier No. PAB10829
Add to Cart
Add to Cart

Specifications

Antigen Goat anti-Rabbit IgG F(ab')2 fragment
Applications Dot Blot, ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Description Goat anti-rabbit IgG recognizes rabbit IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Alkaline Phosphatase.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM Tris-Cl, 150mM NaCl, 1mM MgCl, 0.1mM ZnCl2, 50% (v/v) glycerol, pH 8.0 (0.01% sodium azide)
Host Species Goat
Immunogen Rabbit IgG F(ab')2 fragment
Purification Method This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Quantity 1 mg
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at 4°C. Do not Freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.