Learn More
Arbor Assays Goat Anti-Rabbit IgG Fc, 100MG
SDP
Supplier:  Arbor Assays A009100MG
Encompass_Preferred
An affinity purified Goat Antibody to Rabbit IgG, Fc region. Supplied in PBS with azide as preservative. Affinity purified on a column with covalently coupled rabbit IgG, Fc attached.
Catalog No. 50-227-8563